Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014233.2 | NP_001014255.1 | 3 - 4 | 564 | 72 | |
XM_006251888.1 | XP_006251950.1 | 3 - 4 | 537 | 72 | ||
GenBank mRNA | BC087604.1 | - | 3 - 4 | 567 | 7312219 | 72 |
FQ225412.1 | - | 3 - 4 | 542 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |