Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014233.2 | NP_001014255.1 | 4 - 5 | 668 | 64 | |
XM_006251888.1 | XP_006251950.1 | 4 - 5 | 641 | 64 | ||
GenBank mRNA | BC087604.1 | - | 4 - 5 | 671 | 7312219 | 64 |
FQ225412.1 | - | 4 - 5 | 646 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |