Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251810.2 | XP_006251872.1 | 127 | 63 | ||
XM_008770527.1 | XP_008768749.1 | 133 | 63 |
Gene Symbol: LOC100911492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251839.2 | XP_006251901.1 | 2 - 3 | 392 | 63 | |
XM_008770622.1 | XP_008768844.1 | 2 - 3 | 392 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |