Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013185.1 | NP_001013203.1 | 12 - 13 | 1548 | 76 | |
XM_006250387.1 | XP_006250449.1 | 12 - 13 | 1631 | 76 | ||
GenBank mRNA | BC091388.1 | - | 12 - 13 | 1548 | 7321446 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |