Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135119.2 | NP_001128591.2 | 1 - 2 | 331 | 126 | |
XM_008769851.1 | XP_008768073.1 | 1 - 2 | 1000 | 126 | ||
XM_008769852.1 | XP_008768074.1 | 1 - 2 | 333 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |