Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107786.1 | NP_001101256.1 | 4 - 5 | 426 | 58 | |
XM_006235142.2 | XP_006235204.1 | 4 - 5 | 2001 | 58 | ||
XM_006235143.2 | XP_006235205.1 | 3 - 4 | 1283 | 58 | ||
XM_006235146.2 | XP_006235208.1 | 3 - 4 | 953 | 58 | ||
XM_006235147.2 | XP_006235209.1 | 3 - 4 | 1056 | 58 | ||
XM_006235148.2 | XP_006235210.1 | 2 - 3 | 607 | 58 | ||
XM_008762270.1 | XP_008760492.1 | 5 - 6 | 1444 | 58 | ||
XM_008762271.1 | XP_008760493.1 | 3 - 4 | 839 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |