Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025653.1 | NP_001020824.1 | 2 - 3 | 657 | 119 | |
XM_006238503.2 | XP_006238565.1 | 2 - 3 | 579 | 119 | ||
GenBank mRNA | BC098685.1 | - | 2 - 3 | 657 | 7385408 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |