Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034913.1 | NP_001030085.1 | 3 - 4 | 534 | 103 | |
GenBank mRNA | BC105913.1 | - | 3 - 4 | 534 | 7385857 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |