Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199253.2 | NP_954862.2 | 10 - 11 | 1872 | 67 | |
GenBank mRNA | AY847775.1 | - | 10 - 11 | 1872 | 67 | |
BC133063.1 | - | 10 - 11 | 1803 | 8371686 | 67 | |
FQ219599.1 | - | 1 - 2 | 100 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |