Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053358.1 | NP_445810.1 | 11 - 12 | 1063 | 90 | |
XM_006238532.2 | XP_006238594.1 | 12 - 13 | 1245 | 90 | ||
XM_006238533.2 | XP_006238595.1 | 12 - 13 | 1220 | 90 | ||
XM_006238534.2 | XP_006238596.1 | 12 - 13 | 1202 | 90 | ||
XM_006238535.2 | XP_006238597.1 | 12 - 13 | 1176 | 90 | ||
XM_006238536.2 | XP_006238598.1 | 11 - 12 | 1162 | 90 | ||
XM_006238537.2 | XP_006238599.1 | 11 - 12 | 1136 | 90 | ||
XM_006238538.2 | XP_006238600.1 | 11 - 12 | 1118 | 90 | ||
XM_006238539.2 | XP_006238601.1 | 9 - 10 | 1102 | 90 | ||
XM_008763902.1 | XP_008762124.1 | 12 - 13 | 2898 | 90 | ||
XM_008763903.1 | XP_008762125.1 | 13 - 14 | 2452 | 90 | ||
XM_008763904.1 | XP_008762126.1 | 12 - 13 | 2871 | 90 | ||
XM_008763905.1 | XP_008762127.1 | 12 - 13 | 2853 | 90 | ||
XM_008763906.1 | XP_008762128.1 | 12 - 13 | 2826 | 90 | ||
XM_008763907.1 | XP_008762129.1 | 11 - 12 | 2813 | 90 | ||
XM_008763908.1 | XP_008762130.1 | 11 - 12 | 2787 | 90 | ||
XM_008763909.1 | XP_008762131.1 | 11 - 12 | 2742 | 90 | ||
GenBank mRNA | AF121893.1 | - | 11 - 12 | 1063 | 90 | |
AY724490.1 | - | 12 - 13 | 844 | 90 | ||
BC107666.1 | - | 11 - 12 | 1047 | 7936459 | 90 | |
FQ230168.1 | - | 6 - 7 | 341 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |