Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177931.2 | NP_808792.1 | 3 - 4 | 396 | 105 | |
XM_006238515.2 | XP_006238577.1 | 4 - 5 | 598 | 105 | ||
XM_006238516.2 | XP_006238578.1 | 5 - 6 | 711 | 105 | ||
XM_006238518.2 | XP_006238580.1 | 4 - 5 | 598 | 105 | ||
GenBank mRNA | AB105378.1 | - | 3 - 4 | 396 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |