Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107959.1 | NP_001101429.1 | 3 - 4 | 648 | 79 | |
XM_006238656.2 | XP_006238718.1 | 3 - 4 | 1464 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |