Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134626.1 | NP_001128098.1 | 5 - 6 | 678 | 68 | |
XM_006238709.2 | XP_006238771.1 | 5 - 6 | 889 | 68 | ||
XM_006238710.2 | XP_006238772.1 | 5 - 6 | 877 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |