Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022239.2 | NP_071575.1 | 5 - 6 | 418 | 102 | |
GenBank mRNA | M94555.1 | - | 5 - 6 | 418 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |