Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109599.1 | NP_001103069.1 | 3 - 4 | 286 | 67 | |
XM_008766341.1 | XP_008764563.1 | 3 - 4 | 2552 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |