Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008326.1 | NP_001008327.1 | 2 - 3 | 379 | 74 | |
XM_006243001.2 | XP_006243063.1 | 2 - 3 | 293 | 74 | ||
GenBank mRNA | BC086420.1 | - | 2 - 3 | 379 | 7300740 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |