Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eda2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257051.2 | XP_006257113.1 | 4 - 5 | 502 | 104 | |
XM_006257052.2 | XP_006257114.1 | 4 - 5 | 502 | 104 | ||
XM_008773299.1 | XP_008771521.1 | 4 - 5 | 502 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |