Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252876.2 | XP_006252938.1 | 6 - 7 | 800 | 58 | |
GenBank mRNA | FQ220757.1 | - | 6 - 7 | 610 | 58 | |
FQ232202.1 | - | 6 - 7 | 609 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |