Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011920.1 | NP_001011920.1 | 6 - 7 | 765 | 131 | |
GenBank mRNA | BC079341.1 | - | 6 - 7 | 765 | 7106652 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |