Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047104.1 | NP_001040569.1 | 1 - 2 | 191 | 70 | |
XM_006252908.2 | XP_006252970.1 | 1 - 2 | 839 | 70 | ||
GenBank mRNA | BC098704.1 | - | 1 - 2 | 191 | 7389633 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |