Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107865.1 | NP_001101335.1 | 9 - 10 | 1149 | 80 | |
XM_006236673.2 | XP_006236735.1 | 10 - 11 | 1458 | 80 | ||
XM_006236674.2 | XP_006236736.1 | 10 - 11 | 1455 | 80 | ||
XM_006236675.2 | XP_006236737.1 | 9 - 10 | 1416 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |