Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057205.2 | NP_476553.1 | 12 - 13 | 964 | 83 | |
XM_006236936.2 | XP_006236998.1 | 11 - 12 | 947 | 83 | ||
GenBank mRNA | AF336829.1 | - | 12 - 13 | 973 | 83 | |
BC081743.1 | - | 12 - 13 | 964 | 7124631 | 83 | |
FQ215637.1 | - | 2 - 3 | 81 | 83 | ||
FQ234407.1 | - | 11 - 12 | 924 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |