Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jagn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044272.1 | NP_001037737.1 | 1 - 2 | 187 | 72 | |
GenBank mRNA | BC098749.1 | - | 1 - 2 | 187 | 7365215 | 72 |
FQ225993.1 | - | 1 - 2 | 191 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |