Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015003.2 | NP_001015003.1 | 4 - 5 | 549 | 86 | |
GenBank mRNA | AY973953.1 | - | 4 - 5 | 536 | 86 | |
BC099779.1 | - | 4 - 5 | 534 | 86 | ||
BC127455.1 | - | 4 - 5 | 541 | 7112865 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |