Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105745.1 | NP_001099215.1 | 4 - 5 | 310 | 108 | |
GenBank mRNA | L23862.1 | - | 4 - 5 | 280 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |