Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173426.1 | NP_001166897.1 | 4 - 5 | 350 | 73 | |
XM_006243274.2 | XP_006243336.1 | 4 - 5 | 2036 | 73 | ||
XM_008766329.1 | XP_008764551.1 | 3 - 4 | 459 | 73 | ||
GenBank mRNA | BC169038.1 | - | 4 - 5 | 350 | 9090232 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |