Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013040.1 | NP_001013058.1 | 9 - 10 | 1151 | 73 | |
XM_006243105.2 | XP_006243167.1 | 9 - 10 | 1728 | 73 | ||
XM_006243106.2 | XP_006243168.1 | 9 - 10 | 1743 | 73 | ||
GenBank mRNA | BC082041.1 | - | 9 - 10 | 1151 | 7131532 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |