Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106544.1 | NP_001100014.1 | 3 - 4 | 321 | 70 | |
GenBank mRNA | FQ216824.1 | - | 3 - 4 | 304 | 70 | |
FQ219919.1 | - | 3 - 4 | 304 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |