Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011959.1 | NP_001011959.1 | 1 - 2 | 341 | 65 | |
XM_006235534.1 | XP_006235596.1 | 2 - 3 | 599 | 65 | ||
GenBank mRNA | BC078934.1 | - | 1 - 2 | 341 | 7108104 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |