Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106581.1 | NP_001100051.1 | 6 - 7 | 781 | 82 | |
XM_006257018.2 | XP_006257080.1 | 5 - 6 | 848 | 82 | ||
XM_006257019.2 | XP_006257081.1 | 5 - 6 | 859 | 82 | ||
XM_006257020.2 | XP_006257082.1 | 5 - 6 | 850 | 82 | ||
GenBank mRNA | BC169078.1 | - | 5 - 6 | 795 | 5598223 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |