Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025747.1 | NP_001020918.1 | 2 - 3 | 198 | 75 | |
XM_006257077.2 | XP_006257139.1 | 4 - 5 | 355 | 75 | ||
GenBank mRNA | BC097987.1 | - | 2 - 3 | 198 | 7444864 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |