Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025747.1 | NP_001020918.1 | 4 - 5 | 316 | 96 | |
XM_006257077.2 | XP_006257139.1 | 6 - 7 | 473 | 96 | ||
GenBank mRNA | BC097987.1 | - | 4 - 5 | 316 | 7444864 | 96 |
FQ211792.1 | - | 4 - 5 | 401 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |