Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106973.1 | NP_001100443.1 | 5 - 6 | 503 | 82 | |
XM_006245755.1 | XP_006245817.1 | 6 - 7 | 842 | 82 | ||
GenBank mRNA | BC168922.1 | - | 5 - 6 | 459 | 7122126 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |