Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108267.1 | NP_001101737.1 | 4 - 5 | 728 | 83 | |
XM_006245818.2 | XP_006245880.1 | 4 - 5 | 1484 | 83 | ||
XM_006245819.2 | XP_006245881.1 | 4 - 5 | 1183 | 83 | ||
XM_006245820.2 | XP_006245882.1 | 4 - 5 | 1620 | 83 | ||
XM_006245822.2 | XP_006245884.1 | 4 - 5 | 2310 | 83 | ||
XM_006245823.2 | XP_006245885.1 | 4 - 5 | 1144 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |