Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01424853_m1
          See other NUBP1 GE Assays ›
          Gene Symbol:
          Nubp1
          Gene Name:
          nucleotide binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 5333378 - 5343949 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01424853_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nubp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009619.1 NP_001009619.1 9 - 10 842 89
          GenBank mRNA BC088221.1 - 9 - 10 842 7321552 89
          FQ214074.1 - 8 - 9 786 89
          FQ218444.1 - 9 - 10 855 89
          FQ220638.1 - 9 - 10 855 89
          FQ222868.1 - 9 - 10 859 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002214.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nubp1

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular iron ion homeostasis
          negative regulation of centrosome duplication
          cell growth
          iron-sulfur cluster assembly
          centrosome localization
          protein localization to cell cortex
          ATP binding
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P163018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P163018 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P163018 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P163018 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92931 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740468 - -
          Affymetrix Rat Gene 1.0 ST Array 10740467 - -
          Affymetrix Rat Gene 1.0 ST Array 10740469 - -
          Affymetrix Rat Gene 1.0 ST Array 10740475 - -
          Affymetrix Rat Gene 1.0 ST Array 10740476 - -
          Affymetrix Rat Gene 1.0 ST Array 10740473 - -
          Affymetrix Rat Gene 1.0 ST Array 10740474 - -
          Affymetrix Rat Gene 1.0 ST Array 10740472 - -
          Affymetrix Rat Gene 1.0 ST Array 10740471 - -
          Affymetrix Rat Gene 1.0 ST Array 10740470 - -
          Affymetrix Rat Gene 1.1 ST Array 10740466 - -
          Rat Genome 230 2.0 Array 1385280_PM_at - -
          Rat Genome 230 2.0 Array 1385280_at - -
          Rat Genome 230 2.0 Array 1371503_PM_at - -
          Rat Genome 230 2.0 Array 1384749_at - -
          Rat Genome 230 2.0 Array 1371503_at - -
          Rat Genome 230 2.0 Array 1384749_PM_at - -
          Affymetrix HT Rat Focus Array 1371503_PM_at - -
          Affymetrix HT Rat Focus Array 1384749_PM_at - -
          Affymetrix HT Rat Focus Array 1371503_at - -
          Affymetrix HT Rat Focus Array 1384749_at - -
          Affymetrix Rat Clariom S Assay PSR1000019919.rn.1 TC1000002214.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000019918.rn.1 TC1000002214.rn.1 RTA TC
          Rat Expression Set 230 1371503_at - -
          Rat Expression Set 230 1371503_PM_at - -
          Rat Expression Set 230 1384749_at - -
          Rat Expression Set 230 1384749_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740466 - -
          Rat Genome U34 Set rc_AA819729_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21135964 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370607 - -
          RatRef-12 Expression BeadChip ILMN_64137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0