Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053753.1 | NP_446205.1 | 4 - 5 | 1261 | 90 | |
GenBank mRNA | BC098661.1 | - | 4 - 5 | 1292 | 7384948 | 90 |
J03734.1 | - | 4 - 5 | 1252 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |