Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108641.1 | NP_001102111.1 | 4 - 5 | 665 | 114 | |
XM_006237044.2 | XP_006237106.1 | 5 - 6 | 530 | 114 | ||
XM_006237045.2 | XP_006237107.1 | 5 - 6 | 705 | 114 | ||
XM_006237046.2 | XP_006237108.1 | 5 - 6 | 530 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |