Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106613.2 | NP_001100083.2 | 5 - 6 | 640 | 132 | |
XM_006236962.2 | XP_006237024.1 | 5 - 6 | 714 | 132 | ||
GenBank mRNA | BC167050.1 | - | 5 - 6 | 558 | 7114402 | 132 |
EV776660.1 | - | 2 - 3 | 119 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |