Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106607.1 | NP_001100077.1 | 5 - 6 | 1373 | 72 | |
GenBank mRNA | BC161970.1 | - | 5 - 6 | 1415 | 9028963 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |