Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170546.1 | NP_001164017.1 | 4 - 5 | 897 | 89 | |
NM_001170547.1 | NP_001164018.1 | 4 - 5 | 871 | 89 | ||
NM_001170548.1 | NP_001164019.1 | 4 - 5 | 875 | 89 | ||
XM_006237049.2 | XP_006237111.1 | 4 - 5 | 1246 | 89 | ||
XM_006237050.2 | XP_006237112.1 | 4 - 5 | 935 | 89 | ||
GenBank mRNA | BC166466.1 | - | 3 - 4 | 843 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |