Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ232114.1 | - | 2 - 3 | 180 | 106 | |
FQ233595.1 | - | 2 - 3 | 180 | 106 | ||
FQ234337.1 | - | 2 - 3 | 180 | 106 | ||
FQ234339.1 | - | 2 - 3 | 180 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |