Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106928.1 | NP_001100398.1 | 9 - 10 | 1246 | 117 | |
XM_006245600.2 | XP_006245662.1 | 14 - 15 | 1978 | 117 | ||
XM_008767367.1 | XP_008765589.1 | 14 - 15 | 2754 | 117 | ||
XM_008767369.1 | XP_008765591.1 | 15 - 16 | 2116 | 117 | ||
XM_008767370.1 | XP_008765592.1 | 14 - 15 | 3014 | 117 | ||
XM_008767371.1 | XP_008765593.1 | 14 - 15 | 2751 | 117 | ||
XM_008767372.1 | XP_008765594.1 | 7 - 8 | 891 | 117 | ||
GenBank mRNA | X13412.1 | - | 4 - 5 | 332 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |