Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106926.1 | NP_001100396.1 | 6 - 7 | 1247 | 84 | |
XM_006245520.2 | XP_006245582.1 | 6 - 7 | 1163 | 84 | ||
XM_006245521.2 | XP_006245583.1 | 6 - 7 | 1021 | 84 | ||
XM_006245522.2 | XP_006245584.1 | 6 - 7 | 1016 | 84 | ||
XM_008767365.1 | XP_008765587.1 | 6 - 7 | 1278 | 84 | ||
GenBank mRNA | BC166559.1 | - | 5 - 6 | 920 | 7312233 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |