Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 7 - 8 | 1011 | 93 | |
XM_006256359.2 | XP_006256421.1 | 7 - 8 | 1062 | 93 | ||
XM_006256360.2 | XP_006256422.1 | 6 - 7 | 858 | 93 | ||
XM_006256361.2 | XP_006256423.1 | 7 - 8 | 1085 | 93 | ||
XM_006256362.2 | XP_006256424.1 | 7 - 8 | 977 | 93 | ||
XM_006256363.1 | XP_006256425.1 | 3 - 4 | 419 | 93 | ||
XM_006256364.1 | XP_006256426.1 | 7 - 8 | 971 | 93 | ||
XM_006256365.1 | XP_006256427.1 | 2 - 3 | 277 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |