Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106380.1 | NP_001099850.1 | 4 - 5 | 315 | 75 | |
GenBank mRNA | BC166733.1 | - | 4 - 5 | 248 | 7936238 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |