Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ212194.1 | - | 463 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |