Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137633.1 | NP_001131105.1 | 4 - 5 | 1249 | 90 | |
XM_006240577.2 | XP_006240639.1 | 3 - 4 | 1257 | 90 | ||
GenBank mRNA | FQ212109.1 | - | 3 - 4 | 1007 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |