Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240542.2 | XP_006240604.1 | 11 - 12 | 1577 | 85 | |
XM_008764811.1 | XP_008763033.1 | 11 - 12 | 1731 | 85 | ||
XM_008764812.1 | XP_008763034.1 | 11 - 12 | 2328 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |