Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040128.1 | NP_001035217.1 | 1 - 2 | 504 | 93 | |
XM_006227834.1 | XP_006227896.1 | 2 - 3 | 666 | 93 | ||
XM_006227837.2 | XP_006227899.1 | 2 - 3 | 1183 | 93 | ||
GenBank mRNA | FQ233028.1 | - | 1 - 2 | 53 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |