Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106194.1 | NP_001099664.1 | 2 - 3 | 263 | 52 | |
NM_001201369.1 | NP_001188298.1 | 3 - 4 | 412 | 52 | ||
GenBank mRNA | FM102597.1 | - | 3 - 4 | 412 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |